missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ MSC 1186

Product Code. 18784444
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18784444 50 mg 50mg
18734524 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18784444 Supplier R&D Systems™ Supplier No. 7927/50
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective SRPK family inhibitor

MSC1186 is a potent and selective inhibitor of serine arginine protein kinases (SRPK) 3, 1 and 2 (IC50 values are 0.6 nM, 3 nM and 81 nM respectively in biochemical assay; EC50 values in NanoBRETassay are 40 nM, 98 nM and 149 nM, respectively). Shows no activity at CLK and DYRK families (IC50 >10 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-[[[2-(6-Chloro-7-fluoro-1H-benzimidazol-2-yl)-4-pyrimidinyl]amino]methyl]-2-pyridinyl]-N-methylmethanesulfonamide
CAS 2871698-23-4
Quantity 50 mg
Product Line Tocris
Target SRPK3/STK23
Molecular Formula C19H17ClFN7O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.