Learn More
R&D Systems™ MS 6105
Description
MS 6105 is a potent and selective lactate dehydrogenase (LDH) Degrader (PROTAC™; for LDHA and LDHB respectively, DC50 values are 38 nM and 74 nM; Dmax is 93% and 86%). Comprises an LDH inhibitor warhead connected by a linker to a VHL ligand. Suppresses growth of several pancreatic cancer cell lines (IC50 range is 12.2 μM - 16.1 μM). Bioavailable in vivo.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 2-(5-(Cyclopropylmethyl)-3-(3-(6-(13-(((S)-1-((2S,4R)-4-hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-13-oxotridecanamido)hex-1-yn-1-yl)phenyl)-4-(4-sulfamoylbenzyl)-1H-pyrazol-1-yl)thiazole-4-ca |
| CAS | 2891709-58-1 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Active Degraders |
| Molecular Formula | C65H81N9O9S3 |
| Purity | 98% |