Learn More
IKA™ MS 3.4 Microtiter Attachment
Product Code. 10378672 Shop All IKA Products
Click to view available options
For Use With (Equipment):
Microtitre Plates
Unit Size:
Each
Product Code. 10378672
Brand: IKA™ 0003426400
Only null left
Add to basket
Edge
Add to basket
Designed for use with a microtiter plate. IKA™ MS 3.4 Microtiter Attachment is compatible with the MS 3 basic and S 3 digital minishakers.
  • Included with minishakers MS 3 basic and S 3 digital
TRUSTED_SUSTAINABILITY
Specifications
Description MS 3.4 Microtiter Attachment
For Use With (Equipment) Microtitre Plates