Learn More
Tocris Bioscience™ MS 15203
Potent and selective GPR171 partial agonist
Brand: Tocris Bioscience™ 6299/25
This item is not returnable.
View return policy
Description
MS 15203 is a potent and selective GPR171 partial agonist (EC50 = 90 nM; 28% inhibition of forskolin activated adenylyl cyclase activity in rat hypothalamic memebranes). Exhibits minimal binding to a panel of ∽70 GPCRs and 10 other receptors (<22% of radioligand displaced from the target receptors). Increases neuronal activity in the paraventricular nucleus. Increases food intake and body weight in mice.Specifications
| 5-[(2-Methyl-1-oxo-2-propen-1-yl)amino]-1,3-benzenedicarboxylic acid | |
| 73912-52-4 | |
| >98% | |
| 25 mg |