Learn More
Tocris Bioscience™ MS 15203
Potent and selective GPR171 partial agonist
Brand:  Tocris Bioscience™ 6299/25
Encompass
Product Code. 15716399
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
MS 15203 is a potent and selective GPR171 partial agonist (EC50 = 90 nM; 28% inhibition of forskolin activated adenylyl cyclase activity in rat hypothalamic memebranes). Exhibits minimal binding to a panel of ∽70 GPCRs and 10 other receptors (<22% of radioligand displaced from the target receptors). Increases neuronal activity in the paraventricular nucleus. Increases food intake and body weight in mice.
Specifications
Specifications
5-[(2-Methyl-1-oxo-2-propen-1-yl)amino]-1,3-benzenedicarboxylic acid
73912-52-4
>98%
25 mg
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.