missing translation for 'onlineSavingsMsg'
Learn More

MRS 2179 tetrasodium salt, Tocris Bioscience™
GREENER_CHOICE

Selective P2Y1 antagonist

Brand:  Tocris Bioscience 0900/10

Encompass

Product Code. 13498123

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

A competitive antagonist at P2Y1 receptors (KB = 100 nM). Selective over P2X1 (IC50 = 1.15 μM), P2X3 (IC50 = 12.9 μM), P2X2, P2X4, P2Y2, P2Y4 and P2Y6 receptors. Inhibits the upregulation of NTPDase1 by ATPγS.
Specifications

Specifications

1454889-37-2
10mg
HLJPLHFSEAJJGC-ZKRIHRHSSA-N
[(2R,3S,5R)-5-[6-(methylamino)purin-9-yl]-2-(phosphonooxymethyl)oxolan-3-yl] dihydrogen phosphate;sodium
131848299
>98%
C11H17N5Na4O9P2
2'-Deoxy-N6-methyladenosine 3', 5'-bisphosphate tetrasodium salt
CNC1=NC=NC2=C1N=CN2C3CC(C(O3)COP(=O)(O)O)OP(=O)(O)O.[Na].[Na].[Na].[Na]
517.19
513.16
MRS 2179 tetrasodium salt
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.