Learn More
Tocris Bioscience™ MRS 1220
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
A potent and highly selective antagonist at the human A3 adenosine receptor (Ki values are 0.65, 305, and 52 nM at hA3, rA1 and rA2A respectively. Displays an IC50 value > 1 μM for inhibition of binding to rat A3 receptors).
Specifications
Specifications
| CAS | 183721-15-5 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C21H14ClN5O2 |
| Formula Weight | 403.83 |
| Physical Form | Solid |
| Product Type | MRS 1220 |