missing translation for 'onlineSavingsMsg'
Learn More

MreB Perturbing Compound A22, TRC

Product Code. 30471361
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471361 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471361 Supplier Toronto Research Chemicals Supplier No. TRCM75012525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material MreB Perturbing Compound A22
CAS 22816-60-0
Molecular Formula C8H8Cl2N2S • HCl
Quantity 25 mg
Synonym A 22, S-(3,4-Dichlorobenzyl)thiouronium Chloride, 2-(3,4-Dichlorobenzyl)-2-thio-pseudourea Monohydrochloride, 2-(3,4-Dichlorobenzyl)-2-thio-pseudourea Hydrochloride, (3,4-Dichlorophenyl)methyl Ester Carbamimidothioic Acid Monohydrochloride, (3,4-Dichlorophenyl)methyl Ester Carbamimidothioic Acid Hydrochloride
InChI Formula InChI=1S/C13H19NO3/c1-9(14(3)10(2)13(16)17)12(15)11-7-5-4-6-8-11/h4-10,12,15H,1-3H3,(H,16,17)/t9-,10+,12+/m0/s1
SMILES C[C@H](N(C)[C@H](C)C(O)=O)[C@@H](O)C1=CC=CC=C1
IUPAC Name (2R)-2-[[(1S,2S)-1-hydroxy-1-phenylpropan-2-yl]-methylamino]propanoic acid
Molecular Weight (g/mol) 235.133646
Formula Weight 237.1365
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.