missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MPPED2 (Human) Recombinant Protein
Human MPPED2 full-length ORF ( AAH31582, 1 a.a. - 294 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH31582 |
---|---|
Gene ID (Entrez) | 744 |
Name | metallophosphoesterase domain containing 2 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16162451
|
Abnova™
H00000744-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16172451
|
Abnova™
H00000744-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
- Sequence: MAHGIPSQGKVTITVDEYSSNPTQAFTHYNINQSRFQPPHVHMVDPIPYDTPKPAGHTRFVCISDTHSRTDGIQMPYGDILLHTGDFTELGLPSEVKKFNDWLGNLPYEYKIVIAGNHELTFDKEFMADLVKQDYYRFPSVSKLKPEDFDNVQSLLTNSIYLQDSEVTVKGFRIYGAPWTPWFNGWGFNLPRGQSLLDKWNLIPEGIDILMTHGPPLGFRDWVPKELQRVGCVELLNTVQRRVRPKLHVFGGIHEGYGIMTDGYTTYINASTCTVSFQPTNPPIIFDLPNPQGS
Specifications
AAH31582 | |
metallophosphoesterase domain containing 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
239FB, C11orf8, D11S302E, FAM1B, Hs.46638, dJ1024C24.1, dJ873F21.1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
744 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MPPED2 | |
GST |