Learn More
CPC (Colder) MPCTM Quick-Disconnect Fittings, Hose Barb Insert, Non-Valved, Polycarbonate
Product Code. 30054052
Click to view available options
Diameter (English) Inner:
1/4 in.
1/8 in.
3/8 in.
Material (Seal):
Buna-N
Silicone
Unit Size:
Pack of 25
Product Code. 30054052
Brand: CPC (Colder) 5011254
Only null left
Add to basket
Add to basket
Connect to a coupling body to quickly create a sterile fluid pathway
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Connect to a female coupling body for easy media transfer. Mix and match termination sizes for added flexibility. 0.375 in. nominal flow path size
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/8 in.
Min. Operating Temperature (Metric) -40°F
Outlet Port Quick Disconnect Insert x Hose Barb
Sterilization Method 121°C
Shape Straight
Material (Seal) Buna-N
Color Purple Tint
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.