Learn More
MP Biomedicals™ Porcine Serum
Product Code. 17571126 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17571126 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17571126 Supplier MP Biomedicals™ Supplier No. 092924149
  • ONLINE EXCLUSIVE PRICE
    36.40€
    34.65€ / 100mL
    Save 1.75€
    4% Off
 Restricted Item
Estimated Shipment: 30-12-2026
Specifications
Color Reddish Brown
Species Porcine
Form Liquid
Product Type Porcine Serum
Quantity 100 mL
Concentration 6.5 to 8.6g/dL
Sterility Sterile
Sterilization Method 0.2 μm filtration
Endotoxin Concentration 1.2EU/mL
For Use With (Application) Media supplement in Virus and mycoplasma production applications, as well as certain specialized applications
Hemoglobin Concentration ≤65mg/dL
Mycoplasma Detected No
pH 7 to 8