Learn More
MP Biomedicals™ Poly-L-Lysine Hydrobromide Attachment Factors: 30,000-70,000 MW
Product Code. 11440812 Shop All MP Biomedical Products
Click to view available options
Quantity:
5 mg
Sterility:
Sterile
Unit Size:
5mg
Product Code. 11440812
Brand: MP Biomedicals™ 0219454305
Facilitate attachment, adherence, and spreading in the culture of adherent cells
Available nonsterile or gamma-irradiated.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Product Type Poly-L-Lysine
Sterility Sterile
Quantity 5 mg
Source Synthetic