Learn More
MP Biomedicals™ Linbro™ Microplate Sealer
Shop All MP Biomedical Products
Click to view available options
No. per Pack:
100
1000
Unit Size:
Pack of 100
Pack of 1000
Description
- Easy application and removal for all microplates for both short- and long-term storage Provide tight seal to minimize evaporation and condensation
- Suitable for use between -16 °C and 38 °C
- Transparent tape and is not pierceable
- Nonsterile
Specifications
Specifications
| Color | Clear |
| Material | Acetate |
| Sterility | Non-sterile |
| No. per Pack | 1000 |
| For Use With (Application) | Used for multi-well plates and tissue culture plates |
| Product Type | Microplate Sealer |
| Temperature Range (Metric) | -16°C to +38°C |