Learn More
MP Biomedicals™ L-Broth Agar, Capsules
For the growth of E.coli and certified for plasmid and other bacterial cultures in liquid medium
Brand:  MP Biomedicals™ 113001231
383.68 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11373209
  • 411.28€ / 1kg
Estimated Shipment: 05-06-2024
Specifications
15° to 30°C
Sterile
1 kg
Agar
6.5