missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Hyaluronidase, Bovine Testes
Shop All MP Biomedical Products
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
Description
Hyaluronidase, [37326-33-3]
- Hyaluronate Glycanohydrolase
- Activity: 300 USP units/mg min.
- Lyophilized powder
Specifications
Specifications
| Concentration | ≥300 U/mg |
| Color | Tan |
| pH | 4.5 to 6 |
| For Use With (Application) | Drug dispersion and delivery |
| Content And Storage | -20°C |
| Source | Bovine testes |
| Quantity | 100 mg |
| Product Type | Hyaluronidase |
| Molecular Weight (g/mol) | ≈55 |
| Inhibitors | Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+ |
| Show More |
Safety and Handling
Product Identifier
- Hyaluronidase
- Danger
- Respiratory sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Hyaluronidase
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction