Learn More
MP Biomedicals™ Hyaluronidase, Bovine Testes
Product Code. 11333929 Shop All MP Biomedical Products
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
Product Code. 11333929
Brand: MP Biomedicals™ 0210074080
Bovine Testes
Hyaluronidase, [37326-33-3]
  • Hyaluronate Glycanohydrolase
  • Activity: 300 USP units/mg min.
  • Lyophilized powder
TRUSTED_SUSTAINABILITY
Specifications
Concentration ≥300 U/mg
Color Tan
pH 4.5 to 6
For Use With (Application) Drug dispersion and delivery
Content And Storage -20°C
Source Bovine testes
Quantity 100 mg
Product Type Hyaluronidase
Molecular Weight (g/mol) ≈55
Inhibitors Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+
Form Lyophilized Powder
Show More Show Less
compliance-icons
Product Identifier
  • Hyaluronidase
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Hyaluronidase