Description
For research use only. Not intended for therapeutic use.
Specifications
Specifications
| Content And Storage | 15° to 30°C |
| Form | Crystalline powder |
| Product Type | Dexamethasone |
| Sterility | Sterile |
| Quantity | 1 mg |