Learn More
MP Biomedicals™ GeneClean™ III Kit
Shop All MP Biomedical Products
Click to view available options
:
GeneClean™ III Kit; No. of Reactions: 600 Preps
Description
- GeneClean™ III Kit allows recovery of labelled DNA fragments with ease
- EZ-Glassmilk™ slurry allows flexibility of reaction size and volume
- EZ-Glassmilk™ is easier to suspend than original Glassmilk™
- The GeneClean™ III kit simplifies the process of purifying DNA into three easy steps: Bind, Wash and Elute
- Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications
Specifications
Specifications
| Concentration | New Wash/Ethanol Concentration must be approx. 50% |
| Content And Storage | Room Temperature |
| Format | Kit |
| For Use With (Application) | Desalting, Elimination of proteins from enzymatic reactions, Removal of primers and unincorporated nucleotides from enzymatic reactions, Separation of linearized from uncut vector and isolation of PCR products away from genomic DNA. |
| No. of Reactions | 600 Preps |
| Product Type | GeneClean III Kit |
| Sample Volume | Up to ∼0.5 mL Gel Slice by Volume or ∼1.0 mL Solution |
| Synonym | DNA Clean-Up Kit |
Safety and Handling
Product Identifier
- GeneClean IIl Kit
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : acetic acid, sodiumiodide
Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.