Learn More
MP Biomedicals™ DOB Medium, Powdered Media
Shop All MP Biomedical Products
Click to view available options
Quantity:
227 g
454 g
1000 g
Unit Size:
1kg
227g
454g
Description
- Use 26.7g powder per liter
- Contents/liter: 1.7g YNB, 5g Ammonium Sulfate, 20g Dextrose
Yeast cell culture, Gene expression
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Product Type | Yeast Media |
| Formulation | YNB 1.7g/L, Ammonium Sulfate 5g/L, Dextrose 20g/L |
| For Use With (Application) | For growth of Saccharomyces cerevisiae |
| Quantity | 227 g |