Learn More
MP Biomedicals™ DNA-Erase™ Decontaminant
Product Code. 11485802 Shop All MP Biomedical Products
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11485802 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11485802 Supplier MP Biomedicals™ Supplier No. 04821805
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Removes DNA contamination
DNA-Erase™ is a non-alkaline, non-corrosive and non-carcinogenic cleansing solution that is highly active against DNA contamination. Presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts. The solution contains a surfactant plus an agent that destroys DNA. By following the simple decontamination instructions, DNA is completely destroyed and removed.
  • Eliminates PCR artifacts from all laboratory surfaces and apparatus, including pipetters
  • Stable, heat-resistant, and noncorrosive
  • Supplied as ready-to-use formula
TRUSTED_SUSTAINABILITY
Specifications
Product Type Decontaminant Solution
Volume (English) 16.9 oz.
Volume (Metric) 500 mL
For Use With (Application) Removal of DNA contamination