missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Wash Solution SEWS-M, Concentrated
Shop All MP Biomedical Products
Click to view available options
Quantity:
12 mL
Unit Size:
12mL
Description
- Concentrated SEWS-M is a wash solution included in the FastDNA and all FastDNA Spin Kits and is used to remove impurities once the DNA is bound to the Binding Matrix.
- Purified DNA is eluted from the Binding Matrix after washing with the SEWS-M solution.
- Concentrated SEWS-M is an EtOH-based wash solution used in extraction of DNA to remove impurities once the DNA is bound to the Binding Matrix.
Specifications
Specifications
| Sample Type | Seeds, Spores, Bacteria (Gram +/-), Animal Tissues, Feces, Food, Fungi, Plants, Roots, Soil, Wastewater, Yeast |
| For Use With (Equipment) | FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1 |
| For Use With (Application) | Sample Preparation, Sample Lysis, Nucleic Acid Isolation and Purification, Molecular Biology, Gene Expression, Microbiological Analysis, Gene Function, Gene Regulation |
| Product Type | Concentrated SEWS-M |
| Quantity | 12 mL |
| Sample Volume | Up to 500 mg Per Prep |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction