missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Collagenase, Type 4
Shop All MP Biomedical Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Pancreatic Islet Isolation Grade. Suggested for pancreatic islet isolation. Prepared to contain lower tryptic activity levels. Activity: ≥160U per mg solid. Unit Definition: One unit liberates peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Dialyzed, lyophilized.
Specifications
Specifications
| Concentration | >160 U/mg |
| Color | Brown |
| For Use With (Application) | For tissue dissociation combined with other Enzymes such as Elastase, Trypsin/Papain |
| Content And Storage | 4°C |
| Source | Clostridium histolyticum |
| Quantity | 100 mg |
| Product Type | Collagenase Type 4 |
| Inhibitors | Cysteine, EDTA or o-phenanthroline, alpha2-macroglobulin - a large plasma glycoprotein, alpha2-macroglobulin, alpha1-antitrypsin |
| Form | Lyophilized Powder |
Safety and Handling
Product Identifier
- collagenase, type 4
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : collagenase
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction