Learn More
MP Biomedicals™ Catalase
Product Code. 11463070 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463070 50 mL 50mL
11473070 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11463070 Supplier MP Biomedicals™ Supplier No. 0210040250
Fungal suspension from Aspergillus niger
Catalase, [9001-05-2]
Density: 1.2g/mL
Activity: 1000 units/ml
TRUSTED_SUSTAINABILITY
Specifications
Concentration ≥1000 U/mL
For Use With (Application) Removal of Peroxides, the generation of Oxygen and in coupled systems, for the determination of metabolites
Content And Storage 4°C
Source Aspergillus niger
Quantity 50 mL
Product Type Catalase
Molecular Weight (g/mol) ≈250
Form Liquid