Learn More
MP Biomedicals™ Carbonic Anhydrase
Product Code. 11447322 Shop All MP Biomedical Products
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
Product Code. 11447322
Brand: MP Biomedicals™ 0215387925
  • Activity 2000units/mg
TRUSTED_SUSTAINABILITY
Specifications
Concentration >2000 U/mg
Color Tan
For Use With (Application) Molecular biology, cell biology
Content And Storage 2°C to 8°C
Source Bovine erythrocytes
Quantity 25 mg
Product Type Carbonic Anhydrase
Molecular Weight (g/mol) 30000
Form Lyophilized Powder
compliance-icons
Product Identifier
  • CARBONIC ANHYDRASE
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : carbonic anhydrase