Learn More
MP Biomedicals™ BCIP/NBT Liq. Substrate Plus, 100mL
Product Code. 17532625 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17532625 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17532625 Supplier MP Biomedicals™ Supplier No. 08980771
  • ONLINE EXCLUSIVE PRICE
    158.00€
    150.15€ / 100mL
    Save 7.85€
    4% Off
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Stabilized chromogen solution with greater sensitivity and produces little or no background staining compared with other alternatives.
TRUSTED_SUSTAINABILITY
Specifications
Chromagen Type BCIP or NBT
Form Liquid
For Use With (Application) For visualizing alkaline phosphatase activity in immunoblotting and immunohistology
Quantity 100 mL
Substrate Type BCIP/NBT Substrate
pH 9.8
Storage Requirements 2° to 8°C
Substrate Quantity 100mL