Learn More
MP Biomedicals™ Basal Medium Eagle Modified Vitamin Solution 100X Without L-Glutamine
Product Code. 17367469 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17367469 100 mL 100mL
17388593 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17367469 Supplier MP Biomedicals™ Supplier No. 091600449
  • ONLINE EXCLUSIVE PRICE
    50.70€
    48.30€ / 100mL
    Save 2.40€
    4% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Basal Medium Eagle Modified Vitamin Solution 100X Without L-Glutamine is for use with Mammalian Cell Culture to provide optimum cell growth.
  • Chemically-defined essential components
  • Lot-to-lot consistency
  • Animal-component free media
  • No proteins, hormones, or other growth factors
  • No biological contamination such as viruses, mycoplasma, or prions
Specifications
Concentration 100 X
Product Type Vitamin Supplement
Without Additives No Glutamine
Quantity 100 mL