Learn More
MP Biomedicals™ Aprotinin, From Bovine Lung
From Bovine Lung
Brand: MP Biomedicals™ 0219115880
713.79 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : CAS : 9087-70-1 Weight : 0.00010kg
Description
Aprotinin, [9087-70-1]
- C284H432N84O79S7
- Lyophilized white powder
Specifications
Inhibition Assay | |
Aprotinin from Bovine Lung | |
100mg | |
4°C |
6511.23 | |
5 to 7 | |
Bovine Lung | |
Lyophilized |
Safety and Handling
- Aprotinin
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.