Learn More
MP Biomedicals™ Aprotinin, From Bovine Lung
From Bovine Lung
Brand: MP Biomedicals™ 0219115825
336.67 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : CAS : 9087-70-1 Weight : 0.00010kg
Description
Aprotinin, [9087-70-1]
- C284H432N84O79S7
- Lyophilized white powder
Specifications
Inhibition Assay | |
Aprotinin from Bovine Lung | |
25mg | |
4°C |
6511.23 | |
5 to 7 | |
Bovine Lung | |
Lyophilized |