Learn More
MP Biomedicals™ Aprotinin
Product Code. 11434932 Shop All MP Biomedical Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
Product Code. 11434932
Brand: MP Biomedicals™ 0219455980
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Cell dissociation reagent isolated from bovine lung. Lyophilized.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 4°C
Form Powder
Product Type Trypsin inhibitor
Sterility Sterile
pH 5 to 7
Quantity 100 mg
Source Bovine lung