Learn More
MP Biomedicals™ Aprotinin
Product Code. 11424932 Shop All MP Biomedical Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
Product Code. 11424932
Brand: MP Biomedicals™ 0219455925
Cell dissociation reagent isolated from bovine lung. Lyophilized.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 4°C
Form Powder
Product Type Trypsin inhibitor
Sterility Sterile
pH 5 to 7
Quantity 25 mg
Source Bovine lung