Learn More
MP Biomedicals™ Antibiotics: Mitomycin
Shop All MP Biomedical Products
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
- Format: Powder, Non-sterile
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Specifications
Specifications
| Content And Storage | 4°C |
| Form | Blue powder |
| Product Type | Antibiotic |
| Quantity | 2 mg |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 2
- Carcinogenicity Category 2
- H300-Fatal if swallowed.
- H314-Causes severe skin burns and eye damage.
- H350i-May cause cancer by inhalation.
- H351-Suspected of causing cancer.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : mitomycin C