missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse XL PDGF-BB, Luminex Performance Assay

Product Code. 18442330
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18442330 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18442330 Supplier R&D Systems™ Supplier No. LMXL9945
  • ONLINE EXCLUSIVE PRICE
    245.00€
    231.00€ / 1 sets
    Save 14.00€
    5% Off
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Species Mouse
Gene ID (Entrez) 5155
Synonym c-sis;IBGC5;PDGF2;PDGF-2;PDGFB;Platelet-derived growth factor subunit B;SIS;SSV, PDGFBB
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.