Learn More
R&D Systems™ Mouse XL IL-21, Luminex™ Performance Assay
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Mouse |
| Gene ID (Entrez) | 59067 |
| Synonym | CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |