Learn More
R&D Systems™ Mouse XL GDF-15, Luminex™ Performance Assay
Product Code. 18472490
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 18472490
Brand: R&D Systems™ LMXL8944
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Species Mouse
Gene ID (Entrez) 9518
Synonym GDF15, growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDF, PDFGDF-15, PLAB, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGF-beta, PTGFBPTGF-beta
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.