Learn More
R&D Systems™ Mouse XL Chitinase 3-like 1, Luminex™ Performance Assay
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Mouse |
| Gene ID (Entrez) | 1116 |
| Synonym | AW208766, Brp39, Chi3l1, Chitinase 3 like 1, gp39, HCgp39, YKL-40 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |