missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse XL CCL3/MIP-1 alpha, Luminex™ Performance Assay

Product Code. 18472050
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18472050 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18472050 Supplier R&D Systems™ Supplier No. LMXL450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Species Mouse
Gene ID (Entrez) 6348
Synonym C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.