Learn More
ACROBiosystems Mouse Transferrin R2 Protein, mouse IgG1 Fc Tag, low endotoxin
low endotoxin
Brand: ACROBiosystems TF2-M5269-1MG
Additional Details : Weight : 0.00001kg
Description
Transferrin receptor protein 2 (TFR2) is a single-pass type II membrane protein with a protease associated (PA) domain, an M28 peptidase domain and a transferrin receptor-like dimerization domain. The mutations in TFR2 gene have been associated with hereditary hemochromatosis type III. Furthermore, TFR2 is involved in the uptake of transferrin-bound iron into cells by endocytosis, although its role is minor compared to transferrin receptor 1.
This protein carries a mouse IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 101.1 kDa. The protein migrates as 116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_056614.3 | |
1 mg | |
Arg 103 - Phe 798 | |
Transferrin R2,TFR2 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
101.1 kDa | |
Mouse TFR2, Mouse Fc Tag (TF2-M5269) is expressed from human 293 cells (HEK293). It contains AA Arg 103 - Phe 798 (Accession # Q9JK x 3-1). | |
-20°C | |
Transferrin R2 | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |