missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Mouse TNF alpha ELISA Kits

Product Code. 10067093
Click to view available options
Quantity:
2 Plate Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10067093

Brand: Thermo Scientific™ Pierce™ EMTNFA2

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Measure levels of mouse tumor necrosis factor alpha (TNFa) in mouse serum and cell culture supernatant samples using 96-well plates.

Assay TNFα (mouse tumor necrosis factor alpha) levels in mouse serum and culture supernatant samples with our Thermo Scientific™ Mouse TNF alpha ELISA Kits.

These mouse TNF alpha ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of tumor necrosis factor alpha (TNF-alpha, TNFα) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. The antibody-pair kit consists of the matched TNFα antibody pairs and TNFα standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant mouse TNFα (mouse tumor necrosis factor alpha)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96 well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 50 to 2450pg/mL

  • Sensitivity: < 9pg/mL

  • Precision: Intra-assay CV < 10%; inter-assay CV < 10%

  • Sample Types: Cell culture supernatant, serum

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with mouse IL-2, IL-3, IL-4, IL-5, IL-6, GM-CSF, IFNα or human TNFα. This ELISA has some cross-reactivity with rabbit and cynomologous monkey TNFα in culture supernatants

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 50 to 2450pg/mL
Assay Sensitivity <9pg/mL
Description Mouse TNF alpha ELISA Kit
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Sample Type Cell Culture Supernatant, Serum
Specificity No significant cross-reaction with mouse IL-2, IL-3, IL-4, IL-5, IL-6, GM-CSF, IFNa or human TNFa. This ELISA has some cross-reactivity with rabbit and cynomologous monkey TNFa in culture supernatants
Sufficient For 2 x 96 microplate wells
Kit Contents TMB Substrate Solution, 2 x 13mL, Streptavidin-HRP Concentrate, 2 x 100 μL, Stop Solution, 2 x 13mL, Standard Diluent, 2 x 12mL, Lyophilized Recombinant Mouse TNF alpha Standard, 4 vials, 30X Wash Buffer, 2 x 50mL, Adhesive Plate Covers, 12 ea, Anti-Mouse TNF alpha Precoated 96-well Strip Plate, 2 ea, Biotinylated Antibody Reagent, 2 x 8mL, HRP Dilution Buffer, 2 x 14mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV <10%; inter-assay CV <10%
Quantity 2 Plate Kit
Sample Volume 50 μL
Target Natural and recombinant mouse TNFa (mouse tumor necrosis factor alpha)
Total Assay Time 3 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.