Learn More
R&D Systems™ Mouse SLC6A3/DAT1 (NP_034150.1) VersaClone cDNA
Specifications
Specifications
| Gene | SLC6A3/DAT1 |
| Accession Number | NP_034150.1 |
| Gene ID (Entrez) | 6531 |
| Gene Alias | DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |