Learn More
R&D Systems™ Mouse Plexin B2 (NP_620088.2) VersaClone cDNA
Specifications
Specifications
| Gene | Plexin B2 |
| Accession Number | NP_620088.2 |
| Gene ID (Entrez) | 23654 |
| Gene Alias | MM1, MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2, PLXNB2 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |