missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse NGFR/TNFRSF16 DuoSet ELISA

Product Code. 30021917
Change view
Click to view available options
Conjugate:
Biotin
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conjugate unitSize
30021917 Biotin 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021917 Supplier R&D Systems™ Supplier No. DY1157

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sufficient for at least 15 96-well plates

The Mouse NGF R/TNFRSF16 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse NGFR/TNFRSF16/p75NTR.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Biotin
Assay Range 31.20 to 2000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 18053
Synonym CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF R, NGF receptor, p75 ICD, p75 NTR, TNFRSF16, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.