missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Lipocalin-2/NGAL DuoSet ELISA
Click to view available options
Quantity:
1 Kit (5 Plates)
Unit Size:
1 set
Description
The Mouse Lipocalin-2/NGAL DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Lipocalin-2/NGAL.
Specifications
Specifications
| Assay Range | 7.81 - 500 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3934 |
| Gene Symbol | Lcn2 |
| Host Species | Mouse |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin |
| Target | Lipocalin-2/NGAL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction