missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-7 Quantikine ELISA Kit

Product Code. 18732244
Change view
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18732244 1 x 96-well Plate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18732244 Supplier R&D Systems™ Supplier No. M7000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse IL-7 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-7 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-7 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 8.3pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
Specificity Natural and Recombinant Mouse IL-7
For Use With (Application) ELISA
Gene ID (Entrez) 16196
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym IL7, IL-7 interleukin-7, interleukin 7, Lymphopoietin-1, PBGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.