Learn More
R&D Systems™ Mouse IL-5 Quantikine ELISA Kit
Product Code. 18791794
Change view
Click to view available options
Quantity:
1 Kit
1 Pack
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18791794 1 Pack 1 set
18732424 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18791794 Supplier R&D Systems™ Supplier No. SM5000
Mouse IL-5 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Mouse IL-5 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-5 in cell culture supernates (50 ul), serum (50 ul).
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 7pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Plate
Sample Type Cell Culture Supernatant, Serum
Specificity Natural and Recombinant Mouse IL-5
Target Species Mouse
For Use With (Application) ELISA
Gene ID (Entrez) 16191
Quantity 1 Pack
Sample Volume Cell Culture Supernates (50uL),Serum (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5 T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I