missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-12 p70 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit (1 Plate)
6 Plates
Unit Size:
1 set
Description
The Mouse IL-12 p70 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse IL-12 p70/IL-12A in cell culture supernates (50 ul), serum (50 ul).
Specifications
Specifications
| Assay Range | 7.80 - 500pg/mL |
| Assay Sensitivity | 2.5pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Serum |
| Specificity | Natural and Recombinant Mouse IL-12 p70 |
| Sufficient For | 6 x 96-well Plates |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction