Learn More
Mouse IgG2a Isotype Control (MOPC-173), DyLight™ 650, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA1193D650
Additional Details : Weight : 0.00900kg
Description
MA1-193-D650 can be used as a negative control in any immunoassay where a DyLight 650-conjugated primary antibody of the mouse IgG2a isotype is used. DyLight 650 has an excitation/emission of 652/672nm.
Mouse IgG2a Control antibody specifically detects Mouse IgG2a in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Mouse IgG2a | |
MOPC-173 | |
DyLight 650 | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
The transplantable plasmacytoma MOPC-173 was induced by intraperitoneal injection of mineral oils into BALB/c mice | |
100 μg | |
Isotype Control (Primary) | |
4° C, do not freeze | |
IgG2a κ |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
Not Applicable | |
Liquid |