missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IFN-beta DuoSet ELISA

Product Code. 30021617
Change view
Click to view available options
Format:
96-well Plates
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
30021617 96-well Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021617 Supplier R&D Systems™ Supplier No. DY823405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

Mouse IFN-beta DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IFN-beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IFN-beta.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Kit
Species Mouse
Binding Target IFN-beta
Format 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 3456
Sample Type Cell Culture Supernatant, Serum, Plasma
Synonym Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.