Learn More
ACROBiosystems Mouse Glypican 3 / GPC3 Protein, Fc Tag
Brand: ACROBiosystems GP3-M5254-100UG
Additional Details : Weight : 0.00001kg
Description
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
This protein carries a human IgG1 Fc tag at the C-terminus. This protein contains a furin-like convertase cleavage site, 354-RQYR-357, and will be partially processed into N and C-terminal fragment with calculated MW of 38.0 kDa and 49.1 kDa respectivelySpecifications
NP_057906 | |
100 μg | |
Gln 25 - Met 557 | |
GPC3,OCI5,Glypican-3,GTR2-2,MXR7,DGSX,SDYS ,SGB,SGBS,SGBS1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
38.0 kDa and 49.1 kDa | |
Mouse Glypican 3, Fc Tag (GP3-M5254) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Met 557 (Accession # Q8CFZ4-1). | |
-20°C | |
Glypican 3 | |
Mouse | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |