Learn More
R&D Systems™ Mouse FCRL6/FcRH6 (NP_001158197) VersaClone cDNA
Product Code. 18710629
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18710629 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18710629 Supplier R&D Systems™ Supplier No. RDC2507
  • ONLINE EXCLUSIVE PRICE
    328.00€
    309.75€ / 10µg
    Save 18.25€
    5% Off
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
  • The Mouse FCRL6/FcRH6 (NP_001158197) VersaClone cDNA clone contains the complete ORF for FCRL6/FcRH6, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The FCRL6/FcRH6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Specifications
Gene mFCRL6/FcRH6
Accession Number NP_001158197
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 343413
Content And Storage Store the unopened product at -20 to -80°C
Species Mouse
Quantity 10 μg