missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse EGF DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
1 set
Description
The Mouse EGF DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse EGF.
Specifications
Specifications
| Quantity | 1 Kit (for 15 Plates) |
| Species | Mouse |
| Conjugate | Biotin |
| Assay Range | 3.91 to 250 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 13645 |
| Synonym | beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG, Urogastrone |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction