missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CXCL10/IP-10/CRG-2 DuoSet ELISA
Click to view available options
Format:
15 96-well Plates
5 96-well Plates
Unit Size:
1 set
Pack of 5
Description
The Mouse CXCL10/IP-10/CRG-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse CXCL10/IP-10/CRG-2.
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Kit |
| Species | Mouse |
| Conjugate | Biotin |
| Assay Range | 62.50 to 4000 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Format | 5 96-well Plates |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction